3-(4-Iodophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115905-100MG 115905-250MG 115905-500MG
102898-724EA 1373.85 USD
102898-724 102898-726 102898-728
3-(4-Iodophenyl)oxetane-3-carboxylic acid ≥95%
3-(4-Iodophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉IO₃
MDL Number: MFCD22628743
CAS Number: 1416323-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR