3-(2-Fluorophenyl)oxetan-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
115895-250MG
115895-500MG
115895-1G
102898-664EA
1512.31
USD
102898-664
102898-666
102898-668
3-(2-Fluorophenyl)oxetan-3-amine hydrochloride ≥95%
3-(2-Fluorophenyl)oxetan-3-amine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁ClFNO |
MDL Number:
MFCD19381726 CAS Number: 1332839-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...