tert-Butyl-2-(aminomethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate ≥95%

Supplier: Matrix Scientific

115860-250MG 115860-1G
102899-768EA 1964.62 USD
102899-768 102899-772
tert-Butyl-2-(aminomethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate ≥95%
tert-Butyl-2-(aminomethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MW: 256.35 g/mol
MDL Number: MFCD22628730
CAS Number: 1403766-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR