3-Boc-amino-3-(3-bromophenyl)oxetane ≥95%
Supplier: Matrix Scientific
115838-250MG
115838-500MG
115838-1G
102898-312EA
1512.31
USD
102898-312
102898-314
102898-316
3-Boc-amino-3-(3-bromophenyl)oxetane ≥95%
3-Boc-amino-3-(3-bromophenyl)oxetane
Hazard: Irritant
Formula:
C₁₄H₁₈BrNO₃ |
MDL Number:
MFCD22628719 CAS Number: 1416323-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...