3-Amino-3-(2-bromophenyl)oxetane hydrochloride ≥95%

Supplier: Matrix Scientific

115826-100MG 115826-250MG 115826-500MG
102898-276EA 1373.85 USD
102898-276 102898-278 102898-280
3-Amino-3-(2-bromophenyl)oxetane hydrochloride ≥95%
3-Amino-3-(2-bromophenyl)oxetane hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO
MDL Number: MFCD22628717
CAS Number: 1416323-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR