6-Phenyl-2-oxaspiro[3.3]heptane-6-carbonitrile ≥95%

Supplier: Matrix Scientific

115820-500MG 115820-1G 115820-5G
102899-722EA 1718.46 USD
102899-722 102899-724 102899-726
6-Phenyl-2-oxaspiro[3.3]heptane-6-carbonitrile ≥95%
6-Phenyl-2-oxaspiro[3.3]heptane-6-carbonitrile

Hazard: Irritant

Formula: C₁₃H₁₃NO
MDL Number: MFCD22575232
CAS Number: 1414513-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR