3-Cyano-3-(dimethyamino)oxetane ≥95%

Supplier: Matrix Scientific

115815-1G 115815-5G 115815-10G
102898-216EA 1787.69 USD
102898-216 102898-218 102898-220
3-Cyano-3-(dimethyamino)oxetane ≥95%
3-Cyano-3-(dimethyamino)oxetane

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD22575228
CAS Number: 1414513-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR