3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

115813-1G 115813-5G 115813-10G
102898-204EA 604.62 USD
102898-204 102898-206 102898-208
3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid ≥95%
3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₂O₄
MDL Number: MFCD22575225
CAS Number: 1414513-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR