C-Azetidin-2-ylmethylamine dihydrochloride ≥95%

Supplier: Matrix Scientific

115808-250MG 115808-500MG 115808-1G
102898-176EA 893.85 USD
102898-176 102898-178 102898-180
C-Azetidin-2-ylmethylamine dihydrochloride ≥95%
C-Azetidin-2-ylmethylamine dihydrochloride

Hazard: Irritant

Formula: C₄H₁₂Cl₂N₂
CAS Number: 1414513-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR