(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3-((tert-butoxycarbonyl)amino)oxetan-3-yl)acetate

115791-500MG 115791-1G 115791-250MG
102897-642EA 1809.23 USD
102897-642 102897-644 102899-674
(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid ethyl ester ≥95%
(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid ethyl ester

Hazard: Irritant

Formula: C₁₂H₂₁NO₅
MW: 259.3 g/mol
MDL Number: MFCD22628756
CAS Number: 1416323-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR