Methyl-6-bromo-3,3-dimethyl-2,3-dihydro-1H-indole-4-carboxylate ≥95%

Supplier: Matrix Scientific

115783-250MG 115783-500MG 115783-1G
102897-526EA 1593.85 USD
102897-526 102897-528 102897-530
Methyl-6-bromo-3,3-dimethyl-2,3-dihydro-1H-indole-4-carboxylate ≥95%
Methyl-6-bromo-3,3-dimethyl-2,3-dihydro-1H-indole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄BrNO₂
MDL Number: MFCD22573392
CAS Number: 1389264-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR