Methyl-3-azabicyclo[3.1.1]heptane-6-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

115781-250MG 115781-500MG 115781-1G
102897-518EA 1718.46 USD
102897-518 102897-520 102897-522
Methyl-3-azabicyclo[3.1.1]heptane-6-carboxylate hydrochloride ≥95%
Methyl-3-azabicyclo[3.1.1]heptane-6-carboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₄ClNO₂
MDL Number: MFCD21647390
CAS Number: 1389264-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR