Ethyl-7-(tert-boc-amino)-9-azabicyclo[3.3.1]nonane-3-carboxylate ≥95%

Supplier: Matrix Scientific

115775-250MG 115775-500MG 115775-1G
102897-486EA 3021.54 USD
102897-486 102897-488 102897-490
Ethyl-7-(tert-boc-amino)-9-azabicyclo[3.3.1]nonane-3-carboxylate ≥95%
Ethyl-7-(tert-boc-amino)-9-azabicyclo[3.3.1]nonane-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₄
MDL Number: MFCD22573386
CAS Number: 1363380-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR