5-(3,3-Difluorocyclobutyl)-2H-pyrazol-3-ylamine ≥95%
Supplier: Matrix Scientific
115769-250MG
115769-500MG
115769-1G
102897-476EA
1593.85
USD
102897-476
102897-478
102897-480
5-(3,3-Difluorocyclobutyl)-2H-pyrazol-3-ylamine ≥95%
5-(3,3-Difluorocyclobutyl)-2H-pyrazol-3-ylamine
Hazard: Irritant
Formula:
C₇H₉F₂N₃ |
MDL Number:
MFCD22573380 CAS Number: 1425931-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...