3-(4-Chlorophenyl)pyrazine-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

115764-500MG 115764-1G
102897-454EA 1318.46 USD
102897-454 102897-456
3-(4-Chlorophenyl)pyrazine-2-carbaldehyde ≥95%
3-(4-Chlorophenyl)pyrazine-2-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₇ClN₂O
MDL Number: MFCD21648492
CAS Number: 1404373-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR