5-(Boc-amino)thiazole-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
5-((tert-Butoxycarbonyl)amino)thiazole-2-carboxylic acid
115756-250MG
115756-500MG
115756-1G
102897-408EA
1318.46
USD
102897-408
102897-410
102897-412
5-(Boc-amino)thiazole-2-carboxylic acid ≥97%
5-(Boc-amino)thiazole-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₂N₂O₄S MW: 244.27 g/mol |
MDL Number:
MFCD22415248 CAS Number: 1389264-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...