4-(Azetidin-3-yl)-2,2-dimethylmorpholine ditrifluoroacetate ≥95%
Supplier: Matrix Scientific
115514-250MG
115514-500MG
102896-580EA
413.85
USD
102896-580
102896-582
4-(Azetidin-3-yl)-2,2-dimethylmorpholine ditrifluoroacetate ≥95%
4-(Azetidin-3-yl)-2,2-dimethylmorpholine ditrifluoroacetate
Hazard: Irritant
Formula:
C₉H₁₈N₂O MW: 170.26 g/mol |
MDL Number:
MFCD27987085 CAS Number: 1523606-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...