1-(Azetidin-3-yl)-3-methylazetidin-3-olditrifluoroacetate ≥95%

Supplier: Matrix Scientific

115363-100MG 115363-250MG 115363-500MG
102895-782EA 824.62 USD
102895-782 102895-784 102895-786
1-(Azetidin-3-yl)-3-methylazetidin-3-olditrifluoroacetate ≥95%
1-(Azetidin-3-yl)-3-methylazetidin-3-olditrifluoroacetate

Hazard: Irritant

Formula: C₁₁H₁₆F₆N₂O₅
MDL Number: MFCD26961269
CAS Number: 1651840-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR