1-(Azetidin-3-yl)-4-fluoropiperidine ditrifluoroacetate ≥95%

Supplier: Matrix Scientific

115362-250MG 115362-500MG 115362-1G
102895-728EA 275.38 USD
102895-728 102895-730 102895-780
1-(Azetidin-3-yl)-4-fluoropiperidine ditrifluoroacetate ≥95%
1-(Azetidin-3-yl)-4-fluoropiperidine ditrifluoroacetate

Hazard: Irritant

Formula: C₈H₁₅FN₂
MW: 158.22 g/mol
MDL Number: MFCD22422277
CAS Number: 1706463-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR