1-(Azetidin-3-yl)-3,3-difluoropyrrolidine ditrifluoroacetate ≥95%

Supplier: Matrix Scientific

115361-500MG 115361-1G 115361-5G
102895-722EA 824.62 USD
102895-722 102895-724 102895-726
1-(Azetidin-3-yl)-3,3-difluoropyrrolidine ditrifluoroacetate ≥95%
1-(Azetidin-3-yl)-3,3-difluoropyrrolidine ditrifluoroacetate

Hazard: Irritant

Formula: C₇H₁₂F₂N₂
MDL Number: MFCD22628910
CAS Number: 1706462-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR