1-(Azetidin-3-yl)azetidin-3-oloxalate ≥95%

Supplier: Matrix Scientific

115360-500MG 115360-1G 115360-5G
102895-716EA 1318.46 USD
102895-716 102895-718 102895-720
1-(Azetidin-3-yl)azetidin-3-oloxalate ≥95%
1-(Azetidin-3-yl)azetidin-3-oloxalate

Hazard: Irritant

Formula: C₆H₁₂N₂O
MDL Number: MFCD26959117
CAS Number: 1523606-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR