Azetidin-1-yl(azetidin-3-yl)methanone trifluoroacetate ≥95%
Supplier: Matrix Scientific
115345-100MG
115345-250MG
115345-500MG
102895-700EA
824.62
USD
102895-700
102895-702
102895-704
Azetidin-1-yl(azetidin-3-yl)methanone trifluoroacetate ≥95%
Azetidin-1-yl(azetidin-3-yl)methanone trifluoroacetate
Hazard: Irritant
Formula:
C₇H₁₂N₂O MW: 140.19 g/mol |
MDL Number:
MFCD22628912 CAS Number: 1706436-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...