1-(Azetidin-3-yl)pyrrolidin-2-one trifluoroacetate ≥95%
Supplier: Matrix Scientific
115308-100MG
115308-250MG
115308-500MG
102895-978EA
824.62
USD
102895-978
102895-980
102895-982
1-(Azetidin-3-yl)pyrrolidin-2-one trifluoroacetate ≥95%
1-(Azetidin-3-yl)pyrrolidin-2-one trifluoroacetate
Hazard: Irritant
Formula:
C₇H₁₂N₂O |
MDL Number:
MFCD22628911 CAS Number: 1706436-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...