(2R)-2-Acetamido-3-(3-hydroxyphenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

115253-1G 115253-500MG
102895-734EA 927.69 USD
102895-734 102899-176
(2R)-2-Acetamido-3-(3-hydroxyphenyl)propanoic acid ≥95%
(2R)-2-Acetamido-3-(3-hydroxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD28023551
CAS Number: 1610380-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR