Ethyl-2-(4-benzyl-1H-1,2,3-triazol-1-yl)acetate ≥95%
Supplier: Matrix Scientific
115231-500MG
115231-1G
102895-654EA
624.62
USD
102895-654
102895-656
Ethyl-2-(4-benzyl-1H-1,2,3-triazol-1-yl)acetate ≥95%
Ethyl-2-(4-benzyl-1H-1,2,3-triazol-1-yl)acetate
Hazard: Irritant
Formula:
C₁₃H₁₅N₃O₂ MW: 245.28 g/mol Melting Pt: 97…99 °C |
MDL Number:
MFCD28023541 CAS Number: 1271724-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...