2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole ≥97%

Supplier: Matrix Scientific

115228-500MG 115228-1G
102895-642EA 624.62 USD
102895-642 102895-644
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole ≥97%
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₃ClF₃NO
MDL Number: MFCD18250485
CAS Number: 1248393-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR