4-Bromo-1-methyl-1H-1,3-benzodiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-methyl-1H-benzo[d]imidazole

115223-1G 115223-500MG
102892-572EA 1410.77 USD
102892-572 102895-634
4-Bromo-1-methyl-1H-1,3-benzodiazole ≥97%
4-Bromo-1-methyl-1H-1,3-benzodiazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.07 g/mol
MDL Number: MFCD18378523
CAS Number: 1353679-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR