4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid ≥95%
Supplier: Matrix Scientific
115156-500MG
115156-1G
102895-494EA
693.85
USD
102895-494
102895-496
4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid ≥95%
4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₈H₁₇N₃O₃ |
MDL Number:
MFCD28023534 CAS Number: 1610377-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...