Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate ≥95%

Supplier: Matrix Scientific

115132-500MG 115132-1G
102891-758EA 693.85 USD
102891-758 102891-760
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate ≥95%
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate

Hazard: Irritant

Formula: C₁₆H₁₆FN₃O₅
MDL Number: MFCD28023527
CAS Number: 1610376-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR