1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥97%

Supplier: Matrix Scientific

115117-500MG 115117-1G
102895-456EA 693.85 USD
102895-456 102898-974
1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥97%
1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃N₅
MDL Number: MFCD28023519
CAS Number: 1610377-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR