4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

115087-500MG 115087-1G
102895-394EA 693.85 USD
102895-394 102895-396
4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥95%
4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀ClN₃O
MDL Number: MFCD28023510
CAS Number: 1610377-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR