2-(4-Methoxyphenyl)-4-oxo-4,5-dihydropyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
115085-500MG
115085-1G
102895-386EA
693.85
USD
102895-386
102895-388
2-(4-Methoxyphenyl)-4-oxo-4,5-dihydropyrazolo[1,5-a]pyrazine-3-carbaldehyde ≥95%
2-(4-Methoxyphenyl)-4-oxo-4,5-dihydropyrazolo[1,5-a]pyrazine-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₁N₃O₃ |
MDL Number:
MFCD28023509 CAS Number: 1610377-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...