Methyl-2,6-dioxo-3-{[2-(trimethylsilyl)ethoxy]methyl}-1,2,3,6-tetrahydropyrimidine-4-carboxylate ≥97%
Supplier: Matrix Scientific
115037-500MG
115037-1G
102898-932EA
836.92
USD
102898-932
102898-934
Methyl-2,6-dioxo-3-{[2-(trimethylsilyl)ethoxy]methyl}-1,2,3,6-tetrahydropyrimidine-4-carboxylate ≥97%
Methyl-2,6-dioxo-3-{[2-(trimethylsilyl)ethoxy]methyl}-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Hazard: Irritant
Formula:
C₁₂H₂₀N₂O₅Si MW: 300.39 g/mol |
MDL Number:
MFCD28023508 CAS Number: 1610377-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...