N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide ≥97%
Supplier: Matrix Scientific
115031-500MG
115031-1G
102899-046EA
624.62
USD
102899-046
102899-048
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide ≥97%
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide
Hazard: Irritant
Formula:
C₁₀H₁₀ClF₃N₂O₂ MW: 282.65 g/mol |
MDL Number:
MFCD28023503 CAS Number: 1610047-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...