Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate ≥97%

Supplier: Matrix Scientific

115026-500MG 115026-1G
102895-228EA 204.62 USD
102895-228 102895-230
Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate ≥97%
Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₃
MW: 285.22 g/mol
MDL Number: MFCD28023499
CAS Number: 773089-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR