Methyl-4-(2-cyano-3-ethoxy-3-oxoprop-1-en-1-yl)benzoate ≥97%

Supplier: Matrix Scientific

115025-500MG 115025-1G
102895-224EA 204.62 USD
102895-224 102895-226
Methyl-4-(2-cyano-3-ethoxy-3-oxoprop-1-en-1-yl)benzoate ≥97%
Methyl-4-(2-cyano-3-ethoxy-3-oxoprop-1-en-1-yl)benzoate

Hazard: Irritant

Formula: C₁₄H₁₃NO₄
MDL Number: MFCD06210203
CAS Number: 357231-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR