Methyl-5-bromo-4-nitro-1H-indazole-3-carboxylate

Supplier: Matrix Scientific

113330-1G 113330-5G
102891-938EA 727.69 USD
102891-938 102891-940
Methyl-5-bromo-4-nitro-1H-indazole-3-carboxylate
Methyl-5-bromo-4-nitro-1H-indazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₆BrN₃O₄
MDL Number: MFCD23106303
CAS Number: 1403767-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR