trans-1-Boc-3-amino-4-fluoropyrrolidine

Supplier: Matrix Scientific

Synonyms: trans-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

113116-1G 113116-5G
102897-720EA 1043.08 USD
102897-720 102897-722
trans-1-Boc-3-amino-4-fluoropyrrolidine
trans-1-Boc-3-amino-4-fluoropyrrolidine

Hazard: Irritant

Formula: C₉H₁₇FN₂O₂
MW: 204.25 g/mol
MDL Number: MFCD18207795
CAS Number: 1363382-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR