3-Aminomethyl-3-(hydroxymethyl)oxetane oxalate

Supplier: Matrix Scientific

113045-1G 113045-5G
102897-582EA 581.54 USD
102897-582 102897-584
3-Aminomethyl-3-(hydroxymethyl)oxetane oxalate
3-Aminomethyl-3-(hydroxymethyl)oxetane oxalate

Hazard: Irritant

MDL Number: MFCD27976571
CAS Number: 1523571-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR