3-(Triphenylphosphoranylidene)-2,5-pyrrolidinedione ≥97%

Supplier: Matrix Scientific

087304-500MG 087304-1G
102853-288EA 172.31 USD
102853-288 102853-290
3-(Triphenylphosphoranylidene)-2,5-pyrrolidinedione ≥97%
3-(Triphenylphosphoranylidene)-2,5-pyrrolidinedione

Hazard: Irritant

Formula: C₂₂H₁₈NO₂P
MW: 359.37 g/mol
Melting Pt: 257…260 °C
MDL Number: MFCD22573512
CAS Number: 28118-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR