Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate ≥97%

Supplier: Matrix Scientific

087282-500MG 087282-1G
102853-200EA 403.08 USD
102853-200 102853-202
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate ≥97%
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate

Hazard: Irritant

Formula: C₁₅H₁₄FNO₃
MW: 275.28 g/mol
Melting Pt: 130…131 °C
MDL Number: MFCD22573519
CAS Number: 1407522-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR