Methyl 3-(3-bromo-4-hydroxyphenyl)-2-(hydroxyimino)-propanoate ≥97%

Supplier: Matrix Scientific

087273-500MG 087273-1G
102853-166EA 803.08 USD
102853-166 102853-168
Methyl 3-(3-bromo-4-hydroxyphenyl)-2-(hydroxyimino)-propanoate ≥97%
Methyl 3-(3-bromo-4-hydroxyphenyl)-2-(hydroxyimino)-propanoate

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₄
MW: 288.095 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD22573517
CAS Number: 1407513-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR