Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate ≥97%

Supplier: Matrix Scientific

087272-500MG 087272-1G
102853-162EA 600 USD
102853-162 102853-164
Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate ≥97%
Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate

Hazard: Irritant

Formula: C₁₇H₁₆BrFO₃
MW: 367.22 g/mol
MDL Number: MFCD22573516
CAS Number: 1407516-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR