3-Iodo-5-methyl-1,2-dihydro-1,6-naphthyridin-2-one ≥97%

Supplier: Matrix Scientific

087265-500MG 087265-1G
102853-134EA 1192.31 USD
102853-134 102853-136
3-Iodo-5-methyl-1,2-dihydro-1,6-naphthyridin-2-one ≥97%
3-Iodo-5-methyl-1,2-dihydro-1,6-naphthyridin-2-one

Hazard: Irritant

Formula: C₉H₇IN₂O
Melting Pt: 260…263 °C
MDL Number: MFCD22573530
CAS Number: 1407532-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR