3-Iodo-2-methoxy-5-methyl-1,6-naphthyridine ≥97%
Supplier: Matrix Scientific
087262-500MG
087262-1G
102853-122EA
1192.31
USD
102853-122
102853-124
3-Iodo-2-methoxy-5-methyl-1,6-naphthyridine ≥97%
3-Iodo-2-methoxy-5-methyl-1,6-naphthyridine
Hazard: Irritant
Formula:
C₁₀H₉IN₂O Melting Pt: 174…176 °C |
MDL Number:
MFCD22573542 CAS Number: 1407532-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...