N-(2-Amino-4-fluoro-6-methylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

087210-500MG 087210-1G
102852-912EA 600 USD
102852-912 102852-914
N-(2-Amino-4-fluoro-6-methylphenyl)acetamide ≥95%
N-(2-Amino-4-fluoro-6-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₁FN₂O
Melting Pt: 144…145 °C
MDL Number: MFCD22573532
CAS Number: 1407516-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR