1-Cyclopropyl-2-(1,3-dioxolan-2-yl)ethanone
Supplier: Matrix Scientific
087157-1G
087157-2G
087157-5G
102852-838EA
795.38
USD
102852-838
102852-840
102852-842
1-Cyclopropyl-2-(1,3-dioxolan-2-yl)ethanone
1-Cyclopropyl-2-(1,3-dioxolan-2-yl)ethanone
Hazard: Irritant
Formula:
C₈H₁₂O₃ |
MDL Number:
MFCD18433509 CAS Number: 691363-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...