(1,3-Dioxolan-2-yl)acetic acid methyl ester
Supplier: Matrix Scientific
087155-1G
087155-2G
087155-5G
102852-826EA
795.38
USD
102852-826
102852-828
102852-830
(1,3-Dioxolan-2-yl)acetic acid methyl ester
(1,3-Dioxolan-2-yl)acetic acid methyl ester
Hazard: Irritant
Formula:
C₆H₁₀O₄ Boiling Pt: 94…96 °C (4 Torr) |
MDL Number:
MFCD18433502 CAS Number: 107599-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...