3-(1,3-Dioxan-2-ylethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

087142-1G 087142-2G 087142-5G
102852-748EA 490.77 USD
102852-748 102852-750 102852-752
3-(1,3-Dioxan-2-ylethyl)benzonitrile ≥97%
3-(1,3-Dioxan-2-ylethyl)benzonitrile

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD08741515
CAS Number: 951890-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR