Ethyl-5-(3-bromo-2-thienyl)-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087124-1G 087124-2G 087124-5G
102852-640EA 490.77 USD
102852-640 102852-642 102852-644
Ethyl-5-(3-bromo-2-thienyl)-5-oxovalerate ≥97%
Ethyl-5-(3-bromo-2-thienyl)-5-oxovalerate

Hazard: Irritant

Formula: C₁₁H₁₃BrO₃S
MDL Number: MFCD07782896
CAS Number: 951889-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR